missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ohaus™ MB27 Basic Moisture Analyzer
OHAUS™ MB27 Basic Moisture Analyzer combines high quality and durable construction into a sleek, compact design.
Brand: Ohaus™ 30251541
This item is not returnable.
View return policy
Description
- With 0.01% 0.001g accuracy, the MB27 Moisture Analyzer is suitable for most applications.
- Halogen heating and dual weight-temperature calibration provide fast, reliable results.
- User-friendly touch-button design for easy operation.

Specifications
MB27 | |
LCD | |
90 g | |
2.800 kg |
280.00 x 165.00 x 127.000 mm | |
RS232 (included) | |
0.177 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction