missing translation for 'onlineSavingsMsg'
Learn More
MBD, 98%, Thermo Scientific Chemicals Product Code.: 11966831

MBD, 98%, Thermo Scientific Chemicals

Product Code. 11966831
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11966831

Brand: Thermo Scientific Acros 413540050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33984-50-8
Molecular Formula C14H12N4O4
Molecular Weight (g/mol) 300.26
InChI Key IENONFJSMWUIQQ-UHFFFAOYSA-N
Synonym 4-4-methoxybenzylamino-7-nitrobenzofurazan, 4-4-methoxybenzylamino-7-nitro-2,1,3-benzoxadiazole, mbd, 7-p-methoxybenzylamino-4-nitrobenz-2-oxa-1,3-diazole, 2,1,3-benzoxadiazol-4-amine, n-4-methoxyphenyl methyl-7-nitro, benzofurazan, 4-p-methoxybenzyl amino-7-nitro, n-4-methoxyphenyl methyl-7-nitro-2,1,3-benzoxadiazol-4-amineShow MoreShow Less
PubChem CID 99555
IUPAC Name N-[(4-methoxyphenyl)methyl]-4-nitro-2,1,3-benzoxadiazol-7-amine
SMILES COC1=CC=C(C=C1)CNC2=CC=C(C3=NON=C23)[N+](=O)[O-]

Specifications

Melting Point 176.0°C to 180.0°C
Color Orange to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 300.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material MBD

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468910 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number413540050,413540010
Lot NumberA0448724 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number413540050,413540010
Lot NumberA0412495 Certificate TypeCertificate of Analysis Date01/04/2022 Catalog Number413540050,413540010
Lot NumberA0431461 Certificate TypeCertificate of Analysis Date21/06/2021 Catalog Number413540050,413540010
Product Identifier
  • MBD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MBD, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.