Learn More
MBL™ Borosilicate Glass Class A Volumetric Flask with Stopper
Complies with ISO 1042 and DIN 12664. MBL™ Borosilicate Glass Class A Volumetric Flask with Stopper are manufactured from chemically resistant borosilicate glass.
POA - 234.00 €
Specifications
Certifications/Compliance | Class A, ISO 1042, DIN 12664, Conformity 'H' certified |
---|---|
Closure Material | Polyethylene |
Description | Blue enamel inscriptions |
Material | Borosilicate Glass |
Graduated | Yes |
Product Code | Brand | Capacity (Metric) | Closure Size | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Height (Metric) | Price | Quantity & Availability | |||
12010733
|
MBL
FRD180 |
1000 mL | 24/29 | 300 mm |
77.90 €
Pack of 2 |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
11910326
|
MBL
FRD172 |
100 mL | 14/23 | 170 mm |
69.30 €
Pack of 5 |
In Stock
Log in to see stock. |
|||
11321925
|
MBL
FRD172 |
100 mL | 14/23 | 170 mm | N/A | ||||
12000713
|
MBL
FRD166 |
10 mL | 90 mm |
57.80 €
Pack of 5 |
Estimated Shipment: 28-02-2025
Log in to see stock. |
||||
11371925
|
MBL
FRD182 |
2000 mL | 29/32 | 370 mm |
68.05 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
12643282
|
MBL
FRD174 |
200 mL | 14/23 | 210 mm |
110.00 €
Pack of 5 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||
12020713
|
MBL
FRD167 |
20 mL | 10/19 | 110 mm |
60.65 €
Pack of 5 |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
12060723
|
MBL
FRD176 |
250 mL | 14/23 | 220 mm |
42.70 €
Pack of 2 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||
12633282
|
MBL
FRD168 |
25 mL | 10/19 | 110 mm |
54.80 €
Pack of 5 |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
11381925
|
MBL
FRD184 |
5 L | 34/35 | 475 mm |
234.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
12653282
|
MBL
FRD178 |
500 mL | 19/26 | 260 mm |
53.65 €
Pack of 2 |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
11900326
|
MBL
FRD170 |
50 mL | 12/21 | 140 mm |
62.15 €
Pack of 5 |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
12623282
|
MBL
FRD164 |
5 mL | 10/19 | 70 mm |
48.05 €
Pack of 5 |
Estimated Shipment: 05-03-2025
Log in to see stock. |
|||
Description
- Finely calibrated to Class A tolerances
- Each flask is laser etched with an individual serial number, batch code and date of manufacture
- Conformity 'H' certified
- Inscriptions in blue enamel
- Supplied with polyethylene stopper
- Batch certificate available to download online
Specifications
Class A, ISO 1042, DIN 12664, Conformity 'H' certified | |
Blue enamel inscriptions | |
Yes |
Polyethylene | |
Borosilicate Glass | |
A |