Learn More
MBL™ Double Ended Borosilicate Glass Stirring Rod
Minimize scratching and increase safety with these glass stirring rods. MBL™ Borosilicate Double Ended Glass Stirring Rods are ideal for a wide range of stirring applications.
37.65 € - 61.65 €
Specifications
Diameter (Metric) | 6 mm |
---|---|
Material | Borosilicate Glass |
Quantity | 10/Pk. |
For Use With (Application) | General stirring applications |
Includes | Rounded button and paddle ends |
Description
- Manufactured from borosilicate glass
- With rounded button and paddle ends
- Button end is useful for crushing clumped powder and solids
- Paddle end helps to circulate sediment when stirring
Specifications
6 mm | |
10/Pk. | |
Rounded button and paddle ends |
Borosilicate Glass | |
General stirring applications |