missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCC950 Na, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
Description
- For in vivo experiments in studies regarding the nueroprotective action of individuals on a ketgenic diet using mouse models to study the effect of imflammasome activity on tumor progression with the inhibition of NLRP3
- Study the response of palcental explants by understadning the cholesterol crystal-mediated IL-1B response
- Soluble in water, DMSO, dimethyl formamide or ethanol

Specifications
Specifications
CAS | 256373-96-3 |
Quantity | 10 mg |
Molecular Formula | C20H23N2NaO5S |
Formula Weight | 426.5 |
Physical Form | Crystalline Solid |
Purity | ≥97% |
Molecular Weight (g/mol) | 426.46 |
Chemical Name or Material | MCC950 Na |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MCC950 Na, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction