missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ McCoy's 5A (Modified) Medium
GREENER_CHOICE

McCoy's 5A (Modified) Medium

Brand:  Gibco™ 26600023

 View more versions of this product

Product Code. 11530646

  • 34.51 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. This medium will support the growth of primary mammalian cells derived from normal bone marrow, skin, spleen, kidney, lung, rat embryos, and other tissues.

Dr. Thomas McCoy originally formulated McCoy's 5A medium as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A contains the reducing agent glutathione, bacto-peptone, and a high level of glucose. This product also includes Dr. Hsu's addition of Hanks' salts to enable use outside a CO2 incubator.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ McCoy's 5A is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ McCoy's 5A product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Gibco™ McCoy's 5A (modified) requires serum supplementation, commonly with 10% Fetal Bovine Serum (FBS). Gibco™ McCoy's 5A (modified) uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 13485 standards, FDA
McCoy's 5A (Modified) Medium, 500mL, 1 X Concentration, Liquid Form, With High Glucose, L-glutamine, Bacto-peptone, Phenol Red, 6.9 to 7.1 pH, 12 Months Shelf Life, 280 to 320mOsm/kg Osmolality, Used for Mammalian Cell Culture
Liquid
High Glucose, Glutamine, Phenol Red, Bacto-peptone
Mammalian cell culture
500 mL
2 to 8°C
1 X
McCoy's 5A (Modified) Medium
No HEPES, No Sodium Pyruvate
6.9 to 7.1
For in-vitro diagnostic use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers