missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ McCoy's 5A (Modified) Medium, GlutaMAX™ Supplement
GREENER_CHOICE

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues.

Brand:  Gibco™ 36600088

 View more versions of this product

Product Code. 11504476

  • 391.00 € / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. This medium will support the growth of primary mammalian cells derived from normal bone marrow, skin, spleen, kidney, lung, rat embryos, and other tissues.

This McCoy's 5A is modified as follows:

With: High Glucose, GlutaMAX™, Bacto-peptone, Phenol Red

Without: Sodium Pyruvate

The complete formulation is available.

Dr. Thomas McCoy originally formulated McCoy's 5A medium as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A contains the reducing agent glutathione, bacto-peptone, and a high level of glucose. This product also includes Dr. Hsu's addition of Hanks' salts to enable use outside a CO2 incubator.

Gibco™ McCoy's 5A (modified) requires serum supplementation, commonly with 10% Fetal Bovine Serum (FBS). Gibco™ McCoy's 5A (modified) uses a sodium bicarbonate buffer system (2.2 g / L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 13485 standards, FDA
Liquid
High Glucose, GlutaMAX, Bacto-peptone, Phenol Red
Mammalian cell culture
6.9 to 7.1
McCoy's 5A (Modified) Medium, 10 x 500mL, Liquid, Phenol Red Indicator, With High Glucose, GlutaMAX™, Bacto-Peptone, Phenol Red, For Mammalian Cell Culture
McCoy's 5A (Modified) Medium
No HEPES, No Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
10 x 500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.