missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ McCoy's 5A (Modified) Medium, GlutaMAX™ Supplement
GREENER_CHOICE

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues.

Brand:  Gibco™ 36600021

 View more versions of this product

Product Code. 11594466

  • 44.55 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. This medium will support the growth of primary mammalian cells derived from normal bone marrow, skin, spleen, kidney, lung, rat embryos, and other tissues.

This McCoy's 5A is modified as follows:
With: High Glucose, GlutaMAX™, Bacto-peptone, Phenol Red
Without: Sodium Pyruvate

The complete formulation is available.

Dr. Thomas McCoy originally formulated McCoy's 5A medium as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A contains the reducing agent glutathione, bacto-peptone, and a high level of glucose. This product also includes Dr. Hsu's addition of Hanks' salts to enable use outside a CO2 incubator.

Gibco™ McCoy's 5A (modified) requires serum supplementation, commonly with 10% Fetal Bovine Serum (FBS). Gibco™ McCoy's 5A (modified) uses a sodium bicarbonate buffer system (2.2 g / L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8° C. Protect from light.
Shipping conditions: Ambient.
Shelf life: 12 months from date of manufacture.
Mammalian Cells
1 X
Liquid
Sterile-filtered
No HEPES, No Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
6.9 to 7.1
500 mL
CO2
Animal Origin-free
Mammalian Cell Culture
McCoy's 5A Medium
High Glucose, GlutaMAX, Phenol Red, Bacto-peptone, Sodium Bicarbonate
Sustainable packaging
280 - 320 mOsm/kg
Gibco™, GlutaMAX™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers