missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ McCoy's 5A (Modified) Medium, HEPES
McCoy's 5A (Modified) Medium, HEPES
Brand: Gibco™ 22330021
This item is not returnable.
View return policy
Description
McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. This medium will support the growth of primary mammalian cells derived from normal bone marrow, skin, spleen, kidney, lung, rat embryos, and other tissues.Dr. Thomas McCoy originally formulated McCoy's 5A medium as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A contains the reducing agent glutathione, bacto-peptone, and a high level of glucose. This product also includes Dr. Hsu's addition of Hanks' salts to enable use outside a CO2 incubator.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ McCoy's 5A is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ McCoy's 5A product made in our Grand Island facility (12330-031). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Gibco™ McCoy's 5A (modified) requires serum supplementation, commonly with 10% Fetal Bovine Serum (FBS). Gibco™ McCoy's 5A (modified) uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
McCoy's 5A (Modified) Medium, 500mL, 1 X Concentration, Liquid Form, With High Glucose, L-glutamine, Bacto-peptone, Phenol Red, HEPES, 7 to 7.4 pH, 12 Months Shelf Life, 285 to 325mOsm/kg Osmolality, Used for Mammalian Cell Culture | |
Liquid | |
High Glucose, Glutamine, Bacto-peptone, Phenol Red, HEPES | |
Mammalian cell culture | |
500 mL |
2 to 8°C | |
1 X | |
McCoy's 5A (Modified) Medium | |
No Sodium Pyruvate | |
7 to 7.4 | |
For in-vitro diagnostic use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction