missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mCherry Monoclonal Antibody (16D7), Pacific Blue™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen M11238
This item is not returnable.
View return policy
Description
MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.Specifications
mCherry | |
Monoclonal | |
Pacific Blue | |
mCherry | |
Full-length mCherry was used as the immunogen | |
50 Tests | |
Primary | |
4° C | |
Liquid | |
Tag |
Flow Cytometry, Immunocytochemistry, Western Blot | |
16D7 | |
PBS with 5mM sodium azide; pH 7.5 | |
Rat | |
Affinity chromatography | |
RUO | |
Tag | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction