missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ MD 100 3-in-1 Photometer: Chlorine, pH, Cyanuric Acid

Provides accurate and reproducible results very quickly. Lovibond MD 100 Photometer uses high quality interference filters with long-life LEDs as a light source without any moving parts in a transparent sample chamber. Other features include ease of operation, ergonomic design, compact dimensions and safe handling.

Brand:  Lovibond™ 278010


Product Code. 15227539

  • 605.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Instrument in a carrying case, three round glass vials with lids, one stirring rod and brush, DPD No. 1 tablets (100 pack), DPD No. 3 tablets (100 pack), Phenol red tablets (100 pack), CyA test tablets (100 pack), 4 x AAA batteries, warranty information, Certificate of compliance, instruction manual
This item is not returnable. View return policy

Description

Description

Provides accurate and reproducible results very quickly. Lovibond MD 100 Photometer uses high quality interference filters with long-life LEDs as a light source without any moving parts in a transparent sample chamber. Other features include ease of operation, ergonomic design, compact dimensions and safe handling.

The calibration and software-based adjustment options mean that the MD 100 is also suitable for use as a testing instrument. The tests are conducted using either Lovibond tablet reagents with long term stability and a guaranteed minimum five or 10 year shelf life, VARIO powder reagents or using liquid reagents.
  • Waterproof (as defined in IP68, 1 hour at 0.1m)
  • Automatic switch-off
  • Real-time clock and date calibration mode
  • Backlit LCD display (on keypress)
  • Factory calibration and user calibration; Reset to factory calibration possible
  • Internal ring memory for 16 data sets
  • Infrared interface for test data transfer
Reagents Tablets
Each tablet is hermetically sealed within an individual aluminum foil pocket, protecting against challenging environmental conditions. This packaging keeps each tablet in perfect condition, right up until the time it is needed by the user.
Specifications
  • Chlorine Range: 0.01 to 6.0mg/L Cl2/0.1 to 10mg/L Cl2
  • pH Range: 6.5 to 8.4
  • Cyanuric Acid Range: 0 to 160mg/L
  • Light source: LED, interference filter (IF) and photosensor in transparent cell chamber
  • Wavelength Accuracy: ±1nm
  • Photometric Accuracy: 3% FS (T = 20°C to 25°C)
  • Photometric Resolution: 0.01 A
  • Temperature: 5° to 40°C
  • Relative Humidity: 30 to 90% (non condensing)
  • Power Supply: 4 AAA batteries
  • Operating Time: approx. 17 hours or 5000 test measurements in continuous mode when display backlight is off
  • Dimensions (L x W x H): 15.5 x 7.5 x 3.5cm
  • Weight: basic unit approx. 260g
Note
Specifications for each type of tablet as well as a ‘Certificate of Analysis’ for each lot is available in the download area at www.lovibond.com.
Specifications

Specifications

MD 100 3-in-1 Photometer: Chlorine, pH and Cyanuric Acid (Tablets)
3% FS
Instrument in a carrying case, three round glass vials with lids, one stirring rod and brush, DPD No. 1 tablets (100 pack), DPD No. 3 tablets (100 pack), Phenol red tablets (100 pack), CyA test tablets (100 pack), 4 x AAA batteries, warranty information, Certificate of compliance, instruction manual
Internal ring memory for 16 data sets
Chlorine: 0.01 to 6.0mg/L Cl2/0.1 to 10mg/L Cl2; pH: 6.5 to 8.4; Cyanuric Acid: 0 to 160mg/L
260 g
Backlit LCD
3.5 cm
15.5 cm
LED
0.01 A
7.5 cm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MD100 CHLORINE-PH-STAB (TABLETS) PHOTOMETER IN CAS .

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P233-Keep container tightly closed.
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, butan-2-ol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.