Learn More
Lovibond™ MD 100 Chlorine Duo
Lovibond MD 100 Chlorine Duo, Applications: Testing in the field, laboratory or plant room, Detection Range: 0 to 6mg/L, 0 to 10mg/L, 0 to 2mg/L, 0.1 to 8mg/L, Display Type: LCD, Filling Solution Type: Reagent
Brand:  Lovibond™ 276020
Product Code. 12651705
  • 549.00 € / Each
Explore more special offers
Description
Description
Lovibond MD 100 Chlorine Duo, Applications: Testing in the field, laboratory or plant room, Detection Range: 0 to 6mg/L, 0 to 10mg/L, 0 to 2mg/L, 0.1 to 8mg/L, Display Type: LCD, Filling Solution Type: Reagent
Specifications
Specifications
MD 100 Chlorine Duo
LCD
5°C to 40°C
4 Micro batteries (AAA), 3 round vials (Glass) with lids, tablets reagents and/or liquid reagents or VARIO powder reagent, guarantee sheet, certificate (COC), instruction manual
155 mm
0 to 6 mg/L, 0 to 10 mg/L, 0 to 2 mg/L, 0.1 to 8 mg/L
Reagent
Testing in the field, laboratory or plant room
5 or 10 Year
75 mm
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H290-May be corrosive to metals.
  • H372-Causes damage to organs.
Precautionary Statement
  • P234-Keep only in original container.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P314-Get medical advice/attention if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.