missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ MD 610 Photometer

Enjoy the mobility of a portable photometer with the features of a modern laboratory photometer. The Lovibond MD 610 Photometer now features a Bluetooth inferface for connection to smart phones and tablets to connect to iOS™ and Android™ app for data management and email delivery. The data management then enables analysis and export as a CSV file or graph via email.

Brand:  Lovibond™ 214025

Product Code. 15564273

  • 1320.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Enjoy the mobility of a portable photometer with the features of a modern laboratory photometer. The Lovibond MD 610 Photometer now features a Bluetooth inferface for connection to smart phones and tablets to connect to iOS™ and Android™app for data management and email delivery. The data management then enables analysis and export as a CSV file or graph via email.

  • Waterproof design makes it ideal for a field instrument, yet it has the functionality and features to be used as a laboratory photometer
  • Unit covers all the important parameters for water analysis from aluminum to Zinc and has over 130 methods pre-programmed 
  • Compatible with Tablet, Liquid and Powder reagents and supplied with vial adapters for 13mm and 16mm tubes, the photometer gives flexibility of choice for many tests 
  • In addition to the pre-programmed methods, up to 10 user defined calibration curves can be programmed if required
  • Large graphic display aids analysis by providing on screen method prompts, information regarding test measurement range and reagent type, and automatic countdown timers for accurate reaction periods 
  • Internal memory is capable of storing up to 1000 results with date, time and sample ID; results can be reviewed at any time, and can be transmitted to Smartphones or Tablets via the Bluetooth connection.
  • With 6 LED light sources, the photometer can measure at peak absorbances for many different methods, ensuring result accuracy and reliability for many applications
  • AquaLX™ app, available free of charge, allows you to transfer measurements to smart phones or tablets via Bluetooth
  • Interfaces: Infra red interface for test data transfer (via IRiM); RJ45 socket for internal updates
  • Acid and solvent resistant touch sensitive keypad; audible feedback via integrated beeper
  • Optics: LEDs, Interference Filters (IF) and photo sensor in transparent sample chamber
  • Memory capacity: 1000 data sets with time, date, sample ID
Specifications

Specifications

Operating time 26 hours continuous or 3500 tests
4 x AA
210 x 95 x 45 mm
Instrument in carrying case, 4 AA batteries, 3 photometer vials (24mm), 3 photometer vials (16mm), 2 vial adapters for 16mm and 13mm tubes, stirring/crushing rod, vial cleaning brush, instruction manual and guarantee sheet.
450 g
CE
Graphic
Photometer
± 1 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.