Learn More
Lovibond™ MD200 2-in-1 photometer kit for copper and pH
MD200 2-in-1 photometer kit for copper and pH with tablet reagents includes tablet reagents 3 round vials with lids carry case batteries and instruction manual
Brand:  Lovibond™ 2872102
Product Code. 12671715
  • 708.00 € / Each
Explore more special offers
Description
Description
Versatile, compact and combination of high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic results in precise measurement results at all times.
Versatile use
The MD 200 photometer can be used in almost all areas of water analysis.
Precise and low maintenance
The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance.
User friendly
The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe.
Reagents
Use long-term stable Lovibond™ reagent tablets (5-10 years guaranteed minimum durability) or Lovibond™ liquid reagents.
Specifications
Specifications
Photometer
LCD
Chlorine, Copper, pH, Cyanuric Acid, M-Alkalinity and Iron Measurement