Learn More
Lovibond™ MD200 3-in-1 photometer kit for chlorine pH and cyanuric acid with tablet reagents
includes tablet reagents, 3 round vials with lids, carry case, batteries and instruction manual
Brand:  Lovibond™ 2860102
Product Code. 12691715
  • 636.00 € / Each
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Versatile, compact and: The benchtop photometer MD 200 combines high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic. The result: precise measurement results at all times - no matter where you use it.
Versatile use
The MD 200 photometer can be used in almost all areas of water analysis.
Precise and low maintenance
The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance.
User friendly
The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe.
Reagents
Use long-term stable Lovibond™ reagent tablets (5-10 years guaranteed minimum durability) or Lovibond™ liquid reagents.
Specifications
Specifications
Photometer
LCD
Chlorine, pH and Cyanuric Acid Measurement
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lovibond™ MD200 3-in-1 photometer kit for chlorine pH and cyanuric acid with tablet reagents >