Learn More
Lovibond™ MD200 3-in-1 photometer kit for chlorine pH and M-alkalinity with liquid reagents
includes liquid and tablet reagents, 3 round vials with lids, carry case, batteries and instruction manual
Brand:  Lovibond™ 2889302
Product Code. 12641725
  • 718.00 € / Each
Explore more special offers
Description
Description
Versatile, compact and: The benchtop photometer MD 200 combines high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic. The result: precise measurement results at all times - no matter where you use it.
Versatile use
The MD 200 photometer can be used in almost all areas of water analysis.
Precise and low maintenance
The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance.
User friendly
The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe.
Reagents
Use long-term stable Lovibond™ reagent tablets (5-10 years guaranteed minimum durability) or Lovibond™ liquid reagents.
Specifications
Specifications
Photometer
LCD
Chlorine, pH and M-Alkalinity Measurement
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • PHOTOMETRE MD200 CL - PH - TA en malette
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P234-Keep only in original container.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : potassium iodide, butan-2-ol, N,N-diethylbenzene-1,4-diammonium sulphate,