missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ MD200 COD Photometer Kit

MD200 COD Photometer Kit for 16mm COD vials includes batteries and carry case

Brand:  Lovibond™ 2892502


Product Code. 12641715

  • 621.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Versatile, compact and combination of high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic results in precise measurement results at all times.

Versatile use

The MD 200 photometer can be used in almost all areas of water analysis.

Precise and low maintenance

The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance.

User friendly

The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe.

Reagents

Use long-term stable Lovibond™ reagent tablets (5-10 years guaranteed minimum durability) or Lovibond™ liquid reagents.

Specifications

Specifications

Photometer
LCD
COD
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MD200 photometer, range mg / l .

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H311-Toxic in contact with skin.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H373-May cause damage to organs.
  • EUH208-Contains . May produce an allergic reaction.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, mercury sulphate, disilver sulphate, potassium dichromate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.