missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDL 100907, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
MDL 100907 is a potent and selective 5-HT2A receptor antagonist (Ki = 0.36 nM, IC50 = 3.3-5.1 nM). MDL 100907 exhibits > 80-fold selectivity for 5-HT2A receptors over other serotonergic receptor subtypes. MDL 100907 is neuroprotective; protects cultured dopaminergic neurons from MPP+ induced toxicity. Antipsychotic agent in vivo.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 373.46 |
Chemical Name or Material | MDL 100907 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'