missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ ME107

Product Code. 15610934
Click to view available options
Size:
7.5
10.5
This item is not returnable. View return policy
This item is not returnable. View return policy

Heavy duty chemical protection

  • High resistance to water based chemicals.
  • Good flexibility and elasticity preventing tears. 
  • Chlorinated to harden and cleanse the surface of the glove
TRUSTED_SUSTAINABILITY

Specifications

Back Material Rubber Latex
Certifications/Compliance EN388:2003/4121, EN374:2003/Virus, EN374:2003/ABCKL
Classification Category III
Cuff Material Natural rubber latex
Description Heavy duty chemical protection
Coating Material Natural Rubber Latex
Color Black
Disposable Reusable
Exterior Finish Smooth
Grade Industrial grade
Hand Specific
Latex Free Yes
Powdered Powder Free
Product Type Immersion Gloves
Size 10.5
Sterility Non-sterile
Style ME107
Thickness (Metric) 1.20 mm
Finger Style full fingers
For Use With (Application) Chemical and Liquid Protection
Glove Cuff Color Black
Liner Material Unflocked
Model ME107
Packaging Type 1 pair in a bag; 12 bags in a carton
Quantity 12 pair
Resistant To Water-based Chemicals
Series ME series
Dimensions (L x W) 610 mm
Length (English) 610 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.