missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ ME86 Manyard Shock-Absorbing Lanyard

Features a unique stretchable design with a special woven shock-absorbing inner core and a heavy tubular outer jacket.

Brand:  Honeywell™ 1013970

Product Code. 12655173

  • 427.00 € / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Unique stretchable design
    • Special woven shock-absorbing inner core
    • Heavy tubular outer jacket
  • A safety clip straps onto the harness to hold the Manyard when not in use
  • Unique warning flag that provides visual proof that a lanyard has been involved in a fall and should be removed from service
  • Reliable visual inspection: the integral res core becomes visible if the outer jacket is damaged or worn
  • Hi-tech Teflon™ protective finish on the outer tubular jacket prolongs service life
  • Lightweight and easy to use
  • Ideal for maintenance on cranes or granties
  • When contracted, the elasticized lanyard reduces the risk of tripping or snagging
  • Shock-absorbing material reduces fall arrest forces
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Snap hook, CS20
> 15 kN,Webbing: > 2200 daN, Snap hook: > 2200 daN, CS20 connector: > 2500daN, Energy absorber: > 500daN
1.5 m
Elasticity: 30%
Polyester fiber, Polyamide
Zinc-plated steel, Webbing: polyamide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.