missing translation for 'onlineSavingsMsg'
Learn More

Thermographic Measurements Ltd Thermax Multi-Change Temperature Indicating Paint

Multi-change timed temperature indicating paint

182.00 € - 319.00 €

Specifications

For Use With (Application) Color changing paint to show heat exposure
Certifications/Compliance ISO 9001
Capacity (Metric) 250 mL
For Use With (Equipment) Insulated vessels, motor components, thermal mapping of equipment
View More Specs

Products 4
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
11741199
View Documents
Thermographic Measurements Ltd
IPMC135THE250ML
Pink
319.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11771199
View Documents
Thermographic Measurements Ltd
IPSC458THE250ML
Green
182.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11781199
View Documents
Thermographic Measurements Ltd
IPSC550THE250ML
Orange
182.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11761199
View Documents
Thermographic Measurements Ltd
IPSC275THE250ML
Blue
182.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Timed Temperature Paint
  • Solvent-based
  • No primer necessary on most surfaces
  • Colorfast in direct sunlight
  • Fast reaction time
  • Superior performance in normal and hostile environments
  • Ideal for thermal surveys
  • Strong clear color contrasts
  • Resistant to oil, steam and water
  • Requires no additives
  • Specifications

    Specifications

    Color changing paint to show heat exposure
    ISO 9001
    250 mL
    Insulated vessels, motor components, thermal mapping of equipment
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Thermographic Measurements Ltd Thermax Multi-Change Temperature Indicating Paint

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.