missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Measuring Scoops

Shaped conveniently for sampling

Brand:  Kartell™ 0106400

 View more versions of this product

Product Code. 12687795

  • 85.80 € / Pack of 6
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Shaped conveniently for sampling

  • Shaped conveniently for sampling
  • Flat bottom with beveled angles for easy cleaning
  • Suitable for foodstuff
Specifications

Specifications

Scoop
Flat
White
141 x 386.6 x 69.4 mm
386.6 mm
Polypropylene
1000 mL
Suitable for foodstuff
141 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.