Learn More
Macherey-Nagel™ Med-Test Combi 10™ VET, Refill Pack
Reliable urinalysis for your pet at the point of care.
Brand: Macherey-Nagel™ 930870
This item is not returnable.
View return policy
Description
- The use of urine test strips is acknowledged as a modern screening method in medical practice.
- Within minutes health status can be obtained.
- Simplifies the decision about further diagnostic and therapeutic action.
Specifications
Urine Test Strip | |
Urinalysis Test: Glucose, Ketones, Protein, Blood, Leukocytes,Nitrite, Specific Gravity, pH, Bilirubin and Urobilinogen | |
Refill Pack | |
100 Strips |