missing translation for 'onlineSavingsMsg'
Learn More

Pal International™ MediPal Alcohol-Free Disinfectant Wipes Soft Pack

Contains 200 wipes. Pal International™ MediPal Alcohol-Free Disinfectant Wipes Soft Pack are suitable for general everyday cleaning and disinfection of non-porous hard surfaces and non-invasive medical devices.

Brand:  Pal International™ S625110MPCE

Product Code. 15154439

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Alcohol-free soft pack
  • Contains 200 cleaning and disinfecting wipes. 
  • Easy to transport and dispense
  • Suitable for general everyday cleaning and disinfection of non-porous hard surfaces and non-invasive medical devices.
  • Effective against MRSA, TB, Acinetobacter, Norovirus and HIV
Specifications

Specifications

Disinfectant Wipe
Single-use
300 x 200 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Medipal Disinfectant Wipes

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.

Precautionary Statement
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers