missing translation for 'onlineSavingsMsg'
Learn More

Liebherr™ Spark-Free interior, Lab Fridge

For secure, accurate and long-lasting, spark-free interior, fridge storage of non temperature-sensitive products, the Liebherr™ Spark-Free Interior, Lab Fridge is designed for general purpose applications with Liebherr build-quality you can rely on.

1683.00 € - 2230.00 €

Specifications

Type Laboratory Refrigerator
Temperature Range 1°C to 15°C
No. of Doors 1
Hinge Side Right, reversible
Capacity (Metric) Shelf 40 kg
View More Specs

Products 2
Product Code Brand Capacity (Metric) Style Price Quantity & Availability  
Product Code Brand Capacity (Metric) Style Price Quantity & Availability  
15202596
View Documents
Liebherr™
COOLSAFE 16460
333 L Tall This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15230469
View Documents
Liebherr™
Lkexv 5400
554 L Tall This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Spark-Free Lab Fridge for storage of solvents and reagents, reach-in, 1-section, width 600mm.
  • Temperature range +1 / +15°C, white exterior, commercial-grade moulded polystyrol interior, fan-assisted refrigeration.
  • Climate class 4, refrigerant R 600a, connection rating 1,0A, 230v/50/1-ph,energy consumption 328 kWh/a
  • This model is intended for the spark-free storage of chemicals where a high level of temperature stability is not required
  • ATEX-certified spark-free interior
  • Analogue controls with digital temperature display in the door.
  • Door lock, reversible hinges
  • 40 kg shelf load
Specifications

Specifications

Laboratory Refrigerator
1
40 kg
40 mm HDPE
Digital, External
Automatic
220/240 V
1°C to 15°C
Right, reversible
Steel
Polystyrol
White
Forced-air
50 Hz
Videos
SDS
Documents

Documents

Product Certifications