missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Medium 199 (modified) with Hanks' Salts with Glutamine without Sodium Bicarbonate

Medium 199 (modified) with Hanks' Salts with Glutamine without Sodium Bicarbonate

Brand:  MP Biomedicals™ 091023122

Product Code. 11420275

  • 53.55 € / 10 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • H199 Powder
  • Hanks' salts with L-glutamine
  • Without sodium bicarbonate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 0-5C
Powder
1 x 10 L
Hanks' salts with L-glutamineWithout sodium bicarbonate
Cell Culture Media
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers