missing translation for 'onlineSavingsMsg'
Learn More

Manutan Medium duty shelving with 30 white economy box, blue/orange

Product Code. 12892049
Encompass_Preferred
Click to view available options
Color:
White
This item is not returnable. View return policy

Product Code. 12892049

Brand: Manutan 860R201

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Single stacked boxes for quick and easy access. Capacity of 10kg per box.
  • Six shelves
  • 30 boxes
  • Shelf capacity UDL: 120kg
  • White box
  • Blue/orange
  • Dimensions (H x W x D): 1600mm x 1525mm x 380mm

Specifications

Color White
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.