missing translation for 'onlineSavingsMsg'
Learn More

Manutan Medium duty tubular starter shelving, grey, 2000mm x 1000mm x 800mm

Product Code. 12887878
Encompass_Preferred
This item is not returnable. View return policy

Product Code. 12887878

Brand: Manutan 316M035A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Economic, multi-purpose shelving with easy 'click-in' system. Ideal for various applications from a simple archive shelving to a complete warehousing operation. Perforated every 62. 5mm for shelf adjustment. Simple assembly - simple 'click-in' shelves. Tubular shelves are equipped with welded hooks for easy connection to uprights with all components powder coated for extra durability.

  • Five shelves
  • Grey uprights
  • Shelf capacity UDL: 230kg
  • Dimensions (H x W x D): 2000mm x 1000mm x 800mm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.