Learn More
OHAUS™ Medium Two Prong Sleeves
OHAUS™ Medium Two Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
Brand: OHAUS™ 30400254
This item is not returnable.
View return policy
Description
- Available in red vinyl and fiberglass
- Fiberglass sleeves are ideal for application above 100°C
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
2 | |
Fiberglass | |
Gray |
Prong Sleeve | |
Medium |