missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Mega Rack™ Clinical Tube Rack
GREENER_CHOICE

Creates space and organization for high throughput labs requiring large sample storage. Fisherbrand™ Mega Rack™ Clinical Tube Rack eliminates the need for multiple tube racks.

Brand:  Fisherbrand™ HS24323E

 View more versions of this product

Product Code. 14811442

  • 43.10 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Fully-assembled, polypropylene
  • Sturdy and light weight material for easy transport
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack and eliminate need for multiple tube racks
  • Autoclavable
Specifications

Specifications

Tube Rack
White
Polypropylene
10mL tubes
337 x 267 x 91mm
267 mm
16 mm
Autoclavable
Tubes (10 mL), Tubes (16 mm)
120
1 Each
337 mm
91 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.