missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micronova™ MegaClean™ Heavy-Duty Cleaner
A non-flammable, heavy duty cleaner and degreaser that removes grease, adhesives and photo-resist stains from most surfaces. Available in 1 or 5 gallon bottles and a 1 quart spray bottle.
267.00 € - 420.00 €
Specifications
Description | Heavy Duty Detergent, Degreaser |
---|---|
For Use With (Equipment) | Mops and Buckets or Wipes and Sponge |
Product Code | Brand | Volume (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (English) | Price | Quantity & Availability | |||||
11932475
|
Micronova™
MC1-G |
1 gal. |
352.00 €
15.14 L |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
13137736
|
Micronova™
MC1-5G |
5 gal. |
420.00 €
19 L |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||||
13157736
|
Micronova™
MC1-Q |
1 qt. |
267.00 €
7.60 L |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||||
Description
The Micronova MegaClean™ Heavy-Duty Cleaner offers an effective alternative to toxic, flammable, solvent-based cleaners, such as acetone and isopropyl alcohol. Filtered to 0.1 microns, this cleaner is ideal for semiconductor, electronics, aerospace and clean-build operations.
- Non-flammable, heavy duty cleanroom degreaser
- Removes grease, adhesives and photo-resist stains from most surfaces
- Ideal for semiconductor, electronics, aerospace and clean-build operations
- Filtered to 0.1 microns
- Available in 1 or 5 gallon refill bottles and a 1 quart spray bottle
- Biodegradable
- Water soluble
- Effective at low concentrations
Specifications
Heavy Duty Detergent, Degreaser | |
Mops and Buckets or Wipes and Sponge |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 2-(2-butoxyethoxy)ethanol, alcohols, C9-11, Ethoxylated