Learn More
Thermo Scientific™ Megafuge 8R Cell Culture Centrifuge Package
Carry out clinical and research applications.
Brand:  Thermo Scientific™ 50144363
6924.45 EUR valid until 2025-06-30
Use promo code "24069" to get your promotional price.
Product Code. 15287539
  • 10653.00 € / Each
Explore more special offers
Includes: Heraeus Megafuge 8R, 230V (Mfr PN: 75007214); TX-150 Rotor (Mfr PN: 75005701); TX-150 Roundbucket (set of 4) (Mfr PN: 75005702); Sealing lids (set of 4) (Mfr PN: 75005707); 50mL con skirted and unskirted adapter (set of 4) (Mfr PN: 75005736); 15mL con adapter (set of 4) (Mfr PN: 75005737)
Description
Description
Features a smart, simple interface and exceptional capacity in a compact design.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Megafuge 8R Cell Culture Centrifuge Package
Megafuge 8R, 230V (Mfr PN: 75007214); TX-150 Rotor (Mfr PN: 75005701); TX-150 Roundbucket (set of 4) (Mfr PN: 75005702); Sealing lids (set of 4) (Mfr PN: 75005707); 50mL con skirted and unskirted adapter (set of 4) (Mfr PN: 75005736); 15mL con adapter (set of 4) (Mfr PN: 75005737)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products