Learn More
Applied Biosystems™ Megaplex™ Primer Pools, Human Pools B v3.0
GREENER_CHOICE
Megaplex™ Primer Pools, Human Pools B v3.0
Brand:  Applied Biosystems™ 4444749
Product Code. 11670089
  • 1354.00 € / 1 set
Explore more special offers
Description
Description
Megaplex™ Primer Pools, Human Pools B v3.0 is available under a single part number containing both the Megaplex™ RT Primers and Megaplex™ PreAmp Primers, Human Pools B v3.0. Megaplex™ RT Primers, Human Pool B v3.0 contains RT primers for 377 unique microRNAs and 4 controls. For use when assay sensitivity of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. The microRNA targets represented in this pool tend be less well characterized, more narrowly expressed and⁄or expressed at a lower level compared to Human Pool A. Megaplex™ Primer Pools, Human Pools B v3.0 are intended for use with the TaqMan™ Array Human MicroRNA B Card v3.0, however, they can also be used to prepare cDNA for real-time analysis using the individual TaqMan™ MicroRNA Assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at -20°C
qPCR
RT Primers, PreAmp Primers
Primer Pool
Human
miRNA analysis
Low
Sustainable packaging
Megaplex™
50 reactions
50 Reactions
50 Reactions