missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEGAscript™ SP6 Transcription Kit
Click to view available options
Product Type:
SP6 In Vitro Transcription kit
Quantity:
40 Reactions
Unit Size:
Each
Description
Novel transcription reaction conditions and Ambion's patented, high-yield technology allow the synthesis of 10 to 50 times the amount of RNA produced by conventional transcription reactions. Each Ambion kit includes sufficient reagents for 40 reactions.
- Exclusive, ultrahigh yield technology is fast: 2 hour reaction time
- Amplifies aRNA for gene array analysis and other applications
- Efficiently incorporates many modified nucleotides
- A typical 20μL MEGAscript reaction with 1μg of the pTRI-Xef-1α control template will yield over 100μg of transcript
- Versatility of the MEGAscript kit allows for manipulation of the reactions to include specialized reagents such as modified nucleotides, cap analog, or additional polymerase
- Accessory Products:
- MessageAmp™ aRNA Amplification Kits are powered by MEGAscript technology and have been developed especially for the amplification of cRNA prior to array analysis. To purify in vitro transcription products from free nucleotides, buffer components, and enzymes, use MEGAclear™ Kit (Mfr. No. AM1908) and the MEGAclear-96 Kit (Mfr. No. AM1909).
Specifications
Specifications
| Content And Storage | SP6 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature. |
| Final Product Type | Synthetic mRNA, mRNA |
| Format | Kit |
| Sample Type | mRNA, Synthetic mRNA |
| For Use With (Application) | In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products |
| No. of Reactions | 40 Reactions |
| Product Line | Ambion, MEGAscript |
| Product Type | SP6 In Vitro Transcription kit |
| Quantity | 40 Reactions |
| Promoter | SP6 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction