missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Meker Burner with NF Connection for Natural Gas

Ensures an excellent air-gas mixture and better combustion.

Brand:  Fisherbrand™ 4101/22BBSP

Product Code. 11705406

  • 69.10 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • 24mm diameter burner opening
  • 160mm overall height
  • Gas supply regulation by needle valve
  • Nickel plated steel to guarantee a stable and homogeneous flame
  • Consumption of 62L/hr
  • For use with natural gas/methane

Specifications

Specifications

Meker Burner
Consumption of 62L/hr
160 mm
1 Ea.
Natural Gas, Methane
24 mm
NF Connector
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers