Learn More
Fisherbrand™ Meker Burner, Natural Gas
Fitted with a perforated steel head, which gives off multiple small but very hot flames.
42.35 € - 62.15 €
Specifications
Product Type Meker Burner
Gas Type Natural Gas
No. per Pack 1 Ea.
View More Specs
Products 2
Product Code Brand Diameter (Metric) Outer Inlet Height (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Inlet Height (Metric) Price Quantity & Availability  
12882154
View accessory products View Documents
Fisherbrand™
4101/32
30 mm 192 mm
62.15 €
Each
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
12872154
View Documents
Fisherbrand™
4101/22
24 mm 162 mm
42.35 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
Description
Description
  • The hottest flame temperature for all natural gas burners is between 900° and 1000°C at the tip of the blue flame cone
  • Constructed utilizing a heavy pressure diecast base painted in a blue chemically resistant acrylic finish
  • All burners tested for flame quality and leakage prior to packing
  • Tests are carried out at the standard gas pressure for each specific type gas
  • Temperatures dependent on mains pressure and the chemical construction of the gas which varies in different parts of the world
  • Standard mains pressure for natural gas is 20 millibars
  • Not fitted with an air regulator
Specifications
Specifications
Meker Burner
Natural Gas
1 Ea.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Meker Burner, Natural Gas