missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Meker Safety Burner with Tétine Connection, 24mm Opening

Supplied with a flame failure device that shuts off the gas supply to the burner within approximately 20 seconds of the flame being extinguished.

Brand:  Fisherbrand™ 4101/22TCM

 View more versions of this product

Product Code. 11789777

  • 129.00 € / Each
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Safety features all contained within the burner, reducing need for an electrical supply
  • Constructed in brass and nickel plated
  • Top of burner fitted with perforated steel head, which gives off multiple small but very hot flames
  • Not fitted with an air regulator

Specifications

Specifications

Meker Safety Burner
Brass
24 mm
T-Tine Connector
Natural Gas, Methane
Consumption of 62L/hr.
160 mm
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.