missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Melamine Formaldehyde Instrument Trays

Use this practical tray for instruments, tools and sensitive utensils. VITLAB™ Melamine Formaldehyde Instrument Trays are stable and self-supporting with smooth, easy to clean surfaces.

Brand:  VITLAB™ 72298

 View more versions of this product

Product Code. 15781996

  • 162.00 € / Pack of 5
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • White instrument tray
  • Tall shape
  • Rounded corners
  • Fitting lid made of PS, please order separately
Specifications

Specifications

350 x 250 x 40 mm
40 mm
Melamine
250 mm
White
350 mm
Instrument Tray
Practical tray for instruments, tools, and sensitive utensils
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.