Learn More
Brand™ Melamine Formaldehyde Mortar with Pestle
Resistant to boiling and autoclavable (at 121°C).
Brand:  Brand™ 110710
 View more versions of this product
Product Code. 10038210
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • White in color
  • With foot and spout
  • Provided individually
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Mortar and pestle
90mm
150mm
Melamine formaldehyde