missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ Melamine Resin-Coated Base Cover
Designed for protection of safety storage cabinets.
Brand: Asecos™ 31005
89.82 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 2.00000kg
Description
- Base cover
- With adjusting aids
Specifications
Protection | |
536 x 1086 x 88 mm | |
2.5 kg |