missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanin, MP Biomedicals™
113.42 €
Chemical Identifiers
CAS | 8049-97-6 |
---|---|
Molecular Formula | C18H10N2O4 |
Molecular Weight (g/mol) | 318.288 |
InChI Key | XUMBMVFBXHLACL-UHFFFAOYSA-N |
Synonym | melanin, phaeomelanins, d0wq4g, melanin from sepia officinalis, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1; 2,5 .0; 13,17 .0; 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1^ 2,5 .0^ 13,17 .0^ 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone |
PubChem CID | 6325610 |
SMILES | CC1=C2C3=C(C4=CNC5=C(C(=O)C(=O)C(=C45)C3=CN2)C)C(=O)C1=O |
Description
Chemical Identifiers
8049-97-6 | |
318.288 | |
melanin, phaeomelanins, d0wq4g, melanin from sepia officinalis, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1; 2,5 .0; 13,17 .0; 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1^ 2,5 .0^ 13,17 .0^ 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone | |
CC1=C2C3=C(C4=CNC5=C(C(=O)C(=O)C(=C45)C3=CN2)C)C(=O)C1=O |
C18H10N2O4 | |
XUMBMVFBXHLACL-UHFFFAOYSA-N | |
6325610 |
Specifications
8049-97-6 | |
C18H10N2O4 | |
XUMBMVFBXHLACL-UHFFFAOYSA-N | |
318.288 | |
1780 | |
Melanin |
Brown to Black | |
melanin, phaeomelanins, d0wq4g, melanin from sepia officinalis, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1; 2,5 .0; 13,17 .0; 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1^ 2,5 .0^ 13,17 .0^ 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone | |
CC1=C2C3=C(C4=CNC5=C(C(=O)C(=O)C(=C45)C3=CN2)C)C(=O)C1=O | |
6325610 | |
Granular Powder |