missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Melon™ Gel IgG Spin Purification Kit

Product Code. 10239183 Shop All Thermo Scientific Products
Click to view available options
Quantity:
3 mL
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10239183

Brand: Thermo Scientific™ 45206

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rapidly purify human, mouse and rabbit IgG from serum and other sources with this simpler alternative to traditional Protein A and Protein G methods.

Thermo Scientific™ Melon Gel IgG Purification Resins and Kits comprise a simpler and faster alternative to traditional Protein A and Protein G methods for purification of human, mouse and rabbit IgG from serum and other sources.

Melon Gel IgG Purification Kits enable fast, easy and gentle purification of antibodies from serum and other sample types. Antibody purification with Melon Gel may be performed with gravity-flow columns, spin columns or chromatography cartridges. Melon Gel IgG Purification saves time compared to classical Protein A or Protein G methods without compromising the quality of the antibody. Compared to antibody affinity purification with Protein A or Protein G, Melon Gel IgG Purification is simpler, faster and provides greater yields and higher purity for a wider variety of antibodies species.

Highlights:

  • Simple, one-step protocol – IgG Purification with Melon Gel Resin does not require the binding, washing, and multiple elution steps required by Protein A and Protein G

  • Rapid purification – antibody purification from serum is four to six times faster than Protein A or G methods

  • High recovery and purity – antibody recovery from serum for many species is greater than 90% with greater than 80% purity

  • Minimal dilution – reduced need to concentrate antibody before storage or use

  • Robust purification – Melon Gel IgG Purification Kits work with a wide range of antibodies including many that do not purify well on Protein A or Protein G

  • Gentle purification – no harsh antibody elution conditions helps retain IgG activity

  • Reusable support – Melon Gel Resin can be used for multiple antibody purifications

  • Available in various formats – spin columns, purification kits and chromatography cartridges

TRUSTED_SUSTAINABILITY

Specifications

Product Type IgG Purification Kit
Product Line Melon
Column Type Proprietary Resin, Affinity
Format Kit, Microcentrifuge Spin Column
Stationary Phase Proprietary Ligand
Purification Target IgG Antibodies
Description Melon™ Gel IgG Spin Purification Kit, 3mL, Spin Column, Kit, Microcentrifuge Format, Affinity, Proprietary Resin Column Type, Proprietary Ligand Immobilized Phase, Sufficient For IgG Purification From Approximately 25 x 0.1mL Serum Samples
Content And Storage Sufficient For: IgG purification from approx. 25 x 0.1 mL serum samples (total approx. 25 mg IgG from serum)
• Melon Gel Support, 3 mL
• Melon Gel Purification Buffer, 100 mL
• Spin Columns, 0.8 mL, 27 columns
• Microcentrifuge Tubes, 2 mL, 30 tubes

Store at 4°C
Quantity 3 mL

For Research Use Only. Not for use in diagnostic procedures.

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.