missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ascites Conditioning Reagent for Melon™ Gel Monoclonal IgG Purification Kit

Purify mouse monoclonal antibodies from serum, ascites or hybridoma cell culture supernatant with this high-yield, mild monoclonal IgG purification kit.

Brand:  Thermo Scientific™ 45219

Product Code. 11899360

  • 63.00 € / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Ascites Conditioning Reagent
Conditioning Reagent
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Kit
Pierce Ascites Conditioning Reagent
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZG393892 Certificate TypeCertificate of Analysis Date12/07/2024 Catalog Number45219
Lot NumberYG382273 Certificate TypeCertificate of Analysis Date04/08/2023 Catalog Number45219
Lot NumberYA367060 Certificate TypeCertificate of Analysis Date08/02/2023 Catalog Number45219
Lot NumberSPECS Certificate TypeCertificate of Analysis Date08/02/2023 Catalog Number45219
Lot NumberVF292267 Certificate TypeCertificate of Analysis Date27/06/2020 Catalog Number45219
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Ascites Conditioning Reagent for Melon™ Gel Monoclonal IgG Purification Kit > Ascites Conditioning Reagent; 5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.