missing translation for 'onlineSavingsMsg'
Learn More

Melting Point Standard Set, Reagecon™
BIOPHARMA

Product Code. 13532100 Shop All Reagecon Products
Click to view available options
Quantity:
10 g
Packaging:
3g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Packaging 3g
Quantity 10 g
compliance-icons
Product Identifier
  • MELTING POINT STANDARD CAFFEINE 235 et 238°C
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.