missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM Amino Acids Solution (50X)
GREENER_CHOICE

Product Code. p-7076094
Click to view available options
Quantity:
100 mL
20 x 100 mL
Unit Size:
100mL
Pack of 20
This item is not returnable. View return policy

Product Code. 11570386

Brand: Gibco™ 11130036

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MEM Amino Acids Solution (50X)

Gibco™ MEM Amino Acids Solution is used as a growth supplement for cell culture medium, to increase cell growth and viability. Gibco™ MEM Amino Acids Solution is formulated to contain 50X the essential amino acids (except L-glutamine) found in the standard Minimum Essential Medium (MEM). The complete formulation is available.

Product Use
For in vitro diagnostic use.

Dual-site cGMP Manufacturing and Quality System
Gibco™ MEM Amino Acids Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM Amino Acids Solution product made in our Grand Island facility (11130-051). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance FDA, ISO 13485
Content And Storage 2 to 8°C
Description MEM Amino Acids Solution, 50X Concentration, 100mL, Liquid, 0 to 2 pH, 2 to 8°C Storage Temperature, 12 Months Shelf Life, Room temperature and Ambient Shipping Condition, Used for Cell Culture, Mammalian Cell Culture
Concentration 50 X
Form Liquid
Product Type Amino Acid Supplement
For Use With (Application) Mammalian Cell Culture
pH 0 to 2
Quantity 100 mL
Regulatory Status RUO - research use only
compliance-icons
Product Identifier
  • MEM AMINO ACIDS W/O GLUT (50X)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.