Learn More
Gibco™ MEM Vitamin Solution (100X)
GREENER_CHOICE
MEM Vitamin Solution (100X)
Brand:  Gibco™ 11120037
Product Code. 11550386
  • 20.69 € / 100mL
Edge
Explore more special offers
Description
Description
Gibco™ MEM Vitamin Solution is used as a growth supplement for cell culture medium, to increase cell growth and viability. Gibco™ MEM Vitamin Solution is formulated to contain 100x the vitamins found in the standard Minimum Essential Medium (MEM). The complete formulation is available.
Product Use
For in vitro diagnostic use.
Dual-site cGMP Manufacturing and Quality System
Gibco™ MEM Vitamin Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ MEM Vitamin Solution product made in our Grand Island facility (11120-052). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
–5 to –20°C
MEM Vitamin Solution
100 mL
MEM Vitamin Solution, 100mL, Fewer Resources Used, Less Waste, Used as a Growth Supplement for Cell Culture Medium, to Increase Cell Growth and Viability
Mammalian Cell Culture
RUO - research use only