missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Membrane Screw Caps

Product Code. 10415933 Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric):
54 mm
86 mm
Height (Metric):
25 mm
40 mm
Unit Size:
Pack of 2
Pack of 5
This item is not returnable. View return policy

Product Code. 10415933

Brand: Duran™ 1160168

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Maintain sterility of glass laboratory bottles. Duran™ Membrane Screw Caps permit pressure equalization and tight sealing of contents via welded-in PTFE membrane.

  • Polypropylene cap
  • For use with glass laboratory bottles
  • Welded-in PTFEmMembrane permits pressure equalization and tight sealing
  • Blue
  • Autoclavable
  • Reduces risk of contamination
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
For Use With (Application) Duran laboratory glass bottles
Material Polypropylene
Quantity 2Pack
Weight (Metric) .064kg
Height (Metric) 40 mm
Product Type Vented Cap
Diameter (Metric) 86 mm
Closure Type Membrane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.