Learn More
Fisherbrand™ Traceable™ Memory-Loc™ Data Logging Thermohygrometers with Calibration
Safely secure collected information. Data cannot be overwritten or modified.
Brand:  Fisherbrand™ Traceable™ 6455FS
183.65 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Product Code. 16641323
  • 235.00 € / Each
Explore more special offers
Description
Description
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Store up to 30 unique alarm events
  • Smart-Alarm™ with visual/audible alarm continues to alert even if the unit returns to nonalarm conditions
  • Adjustable sampling rate—from 1 min to 12 hrs in 1 min steps
Specifications
Specifications
Thermohygrometer with Barometer and Calibration
1
±0.4°C
0.1% RH
0°C to 65°C
NIST
±3% RH between 5 to 75%, otherwise ±5% RH
0 to 95%, non-condensing
0.1°C