Learn More
Gilac™ Men's 1 L Urinal
Men's 1 L Urinal is graduated, and suitable for hospital use.
Brand:  Gilac™ G101208
Product Code. 17988601
  • 5.70 € / Each
Explore more special offers
Description
Description
  • Suitable for hospital use.
  • Translucent, level visible, with volume graduations.
  • Rounded design and very thick.
  • Not easy to break when dropped.
  • Outstanding durability even in the intensive use cycle.
  • Product is disinfected in a bedpan washer.
  • Researched product ergonomics ensure optimum cleaning.
  • Equipment at room temperature to avoid the cold sensation experienced with stainless steel equivalents.
Specifications
Specifications
Urinal
Polypropylene
Yes
Green
Non-sterile