missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Menthofuran, 95%, Thermo Scientific Chemicals
52.66 € - 204.02 €
Chemical Identifiers
CAS | 494-90-6 |
---|---|
Molecular Formula | C10H14O |
Molecular Weight (g/mol) | 150.22 |
MDL Number | MFCD00041851 |
InChI Key | YGWKXXYGDYYFJU-UHFFFAOYSA-N |
Synonym | menthofuran, menthofurane, 3,9-epoxy-p-mentha-3,8-diene, benzofuran, 4,5,6,7-tetrahydro-3,6-dimethyl, p-mentha-3,8-diene, 3,9-epoxy, 4,5,6,7-tetrahydro-3,6-dimethylbenzofuran, fema no. 3235, 4,5,6,7-tetrahydro-3,6-dimethylcoumarone, r-menthofuran, 3,6-dimethyl-4,5,6,7-tetrahydrobenzo 2,1-b furan |
PubChem CID | 329983 |
ChEBI | CHEBI:50542 |
IUPAC Name | 3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran |
SMILES | CC1CCC2=C(C1)OC=C2C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10722022
|
Thermo Scientific Acros
413560100 |
10 g | Glass bottle |
52.66 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10432815
|
Thermo Scientific Acros
413560500 |
50 g | Glass bottle |
204.02 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
494-90-6 | |
150.22 | |
YGWKXXYGDYYFJU-UHFFFAOYSA-N | |
329983 | |
3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran |
C10H14O | |
MFCD00041851 | |
menthofuran, menthofurane, 3,9-epoxy-p-mentha-3,8-diene, benzofuran, 4,5,6,7-tetrahydro-3,6-dimethyl, p-mentha-3,8-diene, 3,9-epoxy, 4,5,6,7-tetrahydro-3,6-dimethylbenzofuran, fema no. 3235, 4,5,6,7-tetrahydro-3,6-dimethylcoumarone, r-menthofuran, 3,6-dimethyl-4,5,6,7-tetrahydrobenzo 2,1-b furan | |
CHEBI:50542 | |
CC1CCC2=C(C1)OC=C2C |
Specifications
494-90-6 | |
0.9700g/mL | |
76°C | |
94% min. (GC) | |
1.4832 to 1.4852 | |
0.97 | |
Solubility in water: insoluble | |
CC1CCC2=C(C1)OC=C2C | |
150.22 | |
CHEBI:50542 | |
95% | |
Menthofuran |
Yellow | |
87.0°C (15.0 mmHg) | |
Authentic | |
C10H14O | |
MFCD00041851 | |
menthofuran, menthofurane, 3,9-epoxy-p-mentha-3,8-diene, benzofuran, 4,5,6,7-tetrahydro-3,6-dimethyl, p-mentha-3,8-diene, 3,9-epoxy, 4,5,6,7-tetrahydro-3,6-dimethylbenzofuran, fema no. 3235, 4,5,6,7-tetrahydro-3,6-dimethylcoumarone, r-menthofuran, 3,6-dimethyl-4,5,6,7-tetrahydrobenzo 2,1-b furan | |
YGWKXXYGDYYFJU-UHFFFAOYSA-N | |
3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran | |
329983 | |
150.22 | |
Liquid |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-795-5
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Menthofuran
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only